Assay Details
Gene Symbol: Rab4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013019.2 | NP_037151.2 | 3 - 4 | 348 | 120 | |
XM_008772599.1 | XP_008770821.1 | 3 - 4 | 1187 | 120 | ||
XM_008772600.1 | XP_008770822.1 | 3 - 4 | 399 | 120 | ||
GenBank mRNA | BC062016.1 | - | 3 - 4 | 348 | 120 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008772600.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
