Assay Details
Gene Symbol: Gcdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108896.1 | NP_001102366.1 | 9 - 10 | 1106 | 102 | |
GenBank mRNA | FQ228966.1 | - | 9 - 10 | 1024 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
