Assay Details
Gene Symbol: Slc7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017353.1 | NP_059049.1 | 5 - 6 | 1017 | 70 | |
GenBank mRNA | AB015432.1 | - | 5 - 6 | 1017 | 70 | |
U00995.1 | - | 3 - 4 | 342 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
