Assay Details
Gene Symbol: Atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011927.1 | NP_001011927.1 | 3 - 4 | 561 | 69 | |
GenBank mRNA | BC088462.1 | - | 3 - 4 | 561 | 69 | |
FQ213716.1 | - | 3 - 4 | 577 | 69 | ||
FQ215412.1 | - | 2 - 3 | 296 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
