Assay Details
Gene Symbol: Atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011927.1 | NP_001011927.1 | 5 - 6 | 719 | 74 | |
GenBank mRNA | BC088462.1 | - | 5 - 6 | 719 | 74 | |
FQ213716.1 | - | 5 - 6 | 735 | 74 | ||
FQ215412.1 | - | 4 - 5 | 454 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
