Assay Details
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034146.1 | NP_001029318.1 | 13 - 14 | 2289 | 99 | |
XM_008772609.1 | XP_008770831.1 | 13 - 14 | 2622 | 99 | ||
XM_008772610.1 | XP_008770832.1 | 13 - 14 | 2617 | 99 | ||
GenBank mRNA | BC105892.1 | - | 13 - 14 | 2289 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
autophagy
regulation of autophagy
protein deubiquitination
DNA damage response, signal transduction by p53 class mediator
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
p53 binding
RNA binding
cysteine-type endopeptidase activity
thiol-dependent deubiquitinase
transmembrane transporter binding
DNA repair
autophagy
regulation of autophagy
protein deubiquitination
DNA damage response, signal transduction by p53 class mediator
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
p53 binding
RNA binding
cysteine-type endopeptidase activity
thiol-dependent deubiquitinase
transmembrane transporter binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P157029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20519 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P157029 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P157029 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P157029 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1071355 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P206258 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69336 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42411 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86146 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10808375 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808376 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808366 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808367 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808370 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808374 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808371 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808369 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808363 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808368 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808373 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808365 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808364 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808372 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10808362 | - | - |
Rat Genome 230 2.0 Array | 1372150_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398453_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372150_at | - | - |
Rat Genome 230 2.0 Array | 1398453_at | - | - |
Affymetrix HT Rat Focus Array | 1372150_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372150_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900006005.rn.1 | TC1900000720.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900006007.rn.1 | TC1900000720.rn.1 | RTA TC |
Rat Expression Set 230 | 1372150_PM_at | - | - |
Rat Expression Set 230 | 1372150_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808362 | - | - |
Rat Genome U34 Set | rc_AA859327_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21411310 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369663 | - | - |
