Assay Details
Gene Symbol: Nudt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109010.1 | NP_001102480.1 | 1 - 2 | 103 | 112 | |
XM_006245589.2 | XP_006245651.1 | 1 - 2 | 112 | 112 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001109010.1, XM_006245589.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
