Assay Details
Gene Symbol: Ilkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022606.1 | NP_072128.1 | 5 - 6 | 464 | 107 | |
XM_008767296.1 | XP_008765518.1 | 4 - 5 | 2420 | 107 | ||
XM_008767297.1 | XP_008765519.1 | 3 - 4 | 2745 | 107 | ||
GenBank mRNA | AF095927.1 | - | 5 - 6 | 464 | 107 | |
BC062010.1 | - | 5 - 6 | 466 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
