Assay Details
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245422.2 | XP_006245484.1 | 2 - 3 | 273 | 103 | |
XM_006245423.2 | XP_006245485.1 | 2 - 3 | 472 | 103 | ||
XM_006245425.2 | XP_006245487.1 | 2 - 3 | 270 | 103 | ||
XM_006245427.2 | XP_006245489.1 | 2 - 3 | 317 | 103 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
