Assay Details
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245422.2 | XP_006245484.1 | 4 - 5 | 369 | 88 | |
XM_006245423.2 | XP_006245485.1 | 4 - 5 | 568 | 88 | ||
XM_006245425.2 | XP_006245487.1 | 4 - 5 | 366 | 88 | ||
XM_006245427.2 | XP_006245489.1 | 4 - 5 | 413 | 88 | ||
XM_008767274.1 | XP_008765496.1 | 3 - 4 | 339 | 88 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
