Assay Details
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008381.1 | NP_001008382.1 | 4 - 5 | 293 | 96 | |
XM_006245422.2 | XP_006245484.1 | 5 - 6 | 441 | 96 | ||
XM_006245423.2 | XP_006245485.1 | 5 - 6 | 640 | 96 | ||
XM_006245425.2 | XP_006245487.1 | 5 - 6 | 438 | 96 | ||
XM_006245427.2 | XP_006245489.1 | 5 - 6 | 485 | 96 | ||
XM_008767274.1 | XP_008765496.1 | 4 - 5 | 411 | 96 | ||
XM_008767275.1 | XP_008765497.1 | 4 - 5 | 373 | 96 | ||
GenBank mRNA | BC086355.1 | - | 4 - 5 | 293 | 96 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
