Assay Details
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008381.1 | NP_001008382.1 | 6 - 7 | 415 | 98 | |
XM_006245422.2 | XP_006245484.1 | 7 - 8 | 563 | 98 | ||
XM_006245423.2 | XP_006245485.1 | 7 - 8 | 762 | 98 | ||
XM_006245425.2 | XP_006245487.1 | 7 - 8 | 560 | 98 | ||
XM_006245427.2 | XP_006245489.1 | 7 - 8 | 607 | 98 | ||
XM_008767274.1 | XP_008765496.1 | 6 - 7 | 533 | 98 | ||
XM_008767275.1 | XP_008765497.1 | 6 - 7 | 495 | 98 | ||
GenBank mRNA | BC086355.1 | - | 6 - 7 | 415 | 98 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
