Assay Details
Gene Symbol: D2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106926.1 | NP_001100396.1 | 8 - 9 | 1534 | 83 | |
XM_006245520.2 | XP_006245582.1 | 8 - 9 | 1450 | 83 | ||
XM_006245521.2 | XP_006245583.1 | 8 - 9 | 1308 | 83 | ||
XM_006245522.2 | XP_006245584.1 | 8 - 9 | 1303 | 83 | ||
XM_008767365.1 | XP_008765587.1 | 8 - 9 | 1565 | 83 | ||
GenBank mRNA | BC166559.1 | - | 7 - 8 | 1207 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
