Assay Details
Gene Symbol: Dtymk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106925.1 | NP_001100395.1 | 1 - 2 | 356 | 117 | |
XM_006245519.2 | XP_006245581.1 | 1 - 2 | 285 | 117 | ||
XM_008767363.1 | XP_008765585.1 | 1 - 2 | 285 | 117 | ||
GenBank mRNA | BC159428.1 | - | 1 - 2 | 220 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
