Assay Details
Gene Symbol: Slco4c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002024.1 | NP_001002024.1 | 7 - 8 | 1268 | 69 | |
GenBank mRNA | AF454761.1 | - | 7 - 8 | 1268 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
