Assay Details
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035232.1 | NP_001030309.1 | 2 - 3 | 299 | 64 | |
NM_017299.2 | NP_058995.2 | 2 - 3 | 432 | 64 | ||
XM_006256295.2 | XP_006256357.1 | 2 - 3 | 1321 | 64 | ||
XM_006256296.2 | XP_006256358.1 | 2 - 3 | 324 | 64 | ||
XM_006256297.2 | XP_006256359.1 | 2 - 3 | 748 | 64 | ||
XM_006256298.2 | XP_006256360.1 | 2 - 3 | 310 | 64 | ||
GenBank mRNA | AF099009.1 | - | 2 - 3 | 441 | 64 | |
AF099010.1 | - | 2 - 3 | 441 | 64 | ||
AF173642.1 | - | 2 - 3 | 278 | 64 | ||
AY756271.1 | - | 2 - 3 | 278 | 64 | ||
AY756272.1 | - | 2 - 3 | 278 | 64 | ||
AY756273.1 | - | 2 - 3 | 278 | 64 | ||
BC085742.1 | - | 2 - 3 | 299 | 64 | ||
U38180.1 | - | 2 - 3 | 424 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
