Assay Details
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035232.1 | NP_001030309.1 | 1 - 2 | 67 | 70 | |
GenBank mRNA | AF173642.1 | - | 1 - 2 | 46 | 70 | |
AY756271.1 | - | 1 - 2 | 46 | 70 | ||
AY756272.1 | - | 1 - 2 | 46 | 70 | ||
AY756273.1 | - | 1 - 2 | 46 | 70 | ||
BC085742.1 | - | 1 - 2 | 67 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF173642.1, AY756271.1, AY756272.1, AY756273.1, BC085742.1, NM_001035232.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
