Assay Details
Gene Symbol: Tap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032056.3 | NP_114445.2 | 9 - 10 | 1727 | 99 | |
GenBank mRNA | BC091140.1 | - | 8 - 9 | 1660 | 99 | |
KC222910.1 | - | 8 - 9 | 1638 | 99 | ||
KC222911.1 | - | 8 - 9 | 1638 | 99 | ||
X75306.1 | - | 9 - 10 | 1674 | 99 | ||
X75307.1 | - | 9 - 10 | 1674 | 99 |
Gene Symbol: LOC103689996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772777.1 | XP_008770999.1 | 9 - 10 | 1779 | 99 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
