Assay Details
Gene Symbol: Hnrnph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108532.1 | NP_001102002.1 | 130 | 137 | ||
XM_006256376.2 | XP_006256438.1 | 2 - 3 | 1785 | 137 | ||
XM_006256377.2 | XP_006256439.1 | 2 - 3 | 1724 | 137 | ||
XM_006256378.2 | XP_006256440.1 | 2 - 3 | 1704 | 137 | ||
XM_006256379.2 | XP_006256441.1 | 2 - 3 | 2364 | 137 | ||
XM_006256380.2 | XP_006256442.1 | 1 - 2 | 517 | 137 | ||
GenBank mRNA | FQ215209.1 | - | 2 - 3 | 354 | 137 |
Gene Symbol: LOC100910795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751958.3 | XP_003752006.2 | 1 - 2 | 524 | 137 | |
XM_006256396.2 | XP_006256458.1 | 1 - 2 | 524 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
