Assay Details
Gene Symbol: Hnrnph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108532.1 | NP_001102002.1 | 667 | 88 | ||
XM_006256376.2 | XP_006256438.1 | 6 - 7 | 2322 | 88 | ||
XM_006256377.2 | XP_006256439.1 | 6 - 7 | 2261 | 88 | ||
XM_006256378.2 | XP_006256440.1 | 6 - 7 | 2241 | 88 | ||
XM_006256379.2 | XP_006256441.1 | 6 - 7 | 2901 | 88 | ||
XM_006256380.2 | XP_006256442.1 | 5 - 6 | 1009 | 88 | ||
GenBank mRNA | FQ215209.1 | - | 6 - 7 | 891 | 88 |
Gene Symbol: LOC100910795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751958.3 | XP_003752006.2 | 5 - 6 | 1016 | 88 | |
XM_006256396.2 | XP_006256458.1 | 5 - 6 | 1061 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
