Assay Details
Gene Symbol: Hnrnph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108532.1 | NP_001102002.1 | 463 | 91 | ||
XM_006256376.2 | XP_006256438.1 | 4 - 5 | 2118 | 91 | ||
XM_006256377.2 | XP_006256439.1 | 4 - 5 | 2057 | 91 | ||
XM_006256378.2 | XP_006256440.1 | 4 - 5 | 2037 | 91 | ||
XM_006256379.2 | XP_006256441.1 | 4 - 5 | 2697 | 91 | ||
GenBank mRNA | FQ215209.1 | - | 4 - 5 | 687 | 91 |
Gene Symbol: LOC100910795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256396.2 | XP_006256458.1 | 3 - 4 | 857 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
