Assay Details
Gene Symbol: Mtch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100833.1 | NP_001094303.1 | 4 - 5 | 596 | 68 | |
XM_006256184.1 | XP_006256246.1 | 4 - 5 | 618 | 68 | ||
GenBank mRNA | BC158661.1 | - | 4 - 5 | 596 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
