Assay Details
Gene Symbol: Tead3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098216.1 | NP_001091686.1 | 12 - 13 | 1440 | 93 | |
GenBank mRNA | BC085938.1 | - | 12 - 13 | 1393 | 93 | |
BC099077.1 | - | 12 - 13 | 1440 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P164330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105731 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164330 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164330 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P164330 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P231983 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92966 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11498 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10831917 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831915 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831919 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831912 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831910 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831913 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831923 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831916 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831920 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831921 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831914 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831911 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10831909 | - | - |
Rat Genome 230 2.0 Array | 1384371_at | - | - |
Rat Genome 230 2.0 Array | 1384371_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376615_at | - | - |
Rat Genome 230 2.0 Array | 1376615_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384371_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376615_at | - | - |
Affymetrix HT Rat Focus Array | 1376615_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384371_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2000010155.rn.1 | TC2000001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000010156.rn.1 | TC2000001216.rn.1 | RTA TC |
Rat Expression Set 230 | 1384371_at | - | - |
Rat Expression Set 230 | 1376615_PM_at | - | - |
Rat Expression Set 230 | 1384371_PM_at | - | - |
Rat Expression Set 230 | 1376615_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10831909 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22138186 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366257 | - | - |
