Assay Details
Gene Symbol: Tead3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098216.1 | NP_001091686.1 | 7 - 8 | 768 | 97 | |
GenBank mRNA | BC085938.1 | - | 7 - 8 | 721 | 97 | |
BC099077.1 | - | 7 - 8 | 768 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
