Assay Details
Gene Symbol: Tead3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098216.1 | NP_001091686.1 | 10 - 11 | 1143 | 59 | |
GenBank mRNA | BC085938.1 | - | 10 - 11 | 1096 | 59 | |
BC099077.1 | - | 10 - 11 | 1143 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
