Assay Details
Gene Symbol: Glo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207594.2 | NP_997477.1 | 2 - 3 | 241 | 147 | |
GenBank mRNA | BC061570.1 | - | 2 - 3 | 245 | 147 | |
FQ210595.1 | - | 2 - 3 | 219 | 147 | ||
FQ220772.1 | - | 2 - 3 | 241 | 147 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
