Assay Details
Gene Symbol: Dnmt3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003964.1 | NP_001003964.1 | 8 - 9 | 848 | 82 | |
XM_006256250.2 | XP_006256312.1 | 7 - 8 | 871 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
