Assay Details
Gene Symbol: Tap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032055.4 | NP_114444.2 | 5 - 6 | 1239 | 76 | |
GenBank mRNA | KC222905.1 | - | 5 - 6 | 997 | 76 | |
Y10234.1 | - | 5 - 6 | 1180 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
