Assay Details
Gene Symbol: Mdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033235.1 | NP_150238.1 | 8 - 9 | 909 | 70 | |
GenBank mRNA | AF093773.1 | - | 8 - 9 | 909 | 70 | |
BC059124.1 | - | 8 - 9 | 950 | 70 | ||
FQ212702.1 | - | 8 - 9 | 953 | 70 | ||
FQ215129.1 | - | 8 - 9 | 964 | 70 | ||
FQ217383.1 | - | 8 - 9 | 953 | 70 | ||
FQ217819.1 | - | 953 | 70 | |||
FQ224891.1 | - | 8 - 9 | 951 | 70 | ||
FQ233582.1 | - | 8 - 9 | 953 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
