Assay Details
Gene Symbol: Mdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033235.1 | NP_150238.1 | 2 - 3 | 146 | 112 | |
GenBank mRNA | AF093773.1 | - | 2 - 3 | 146 | 112 | |
BC059124.1 | - | 2 - 3 | 187 | 112 | ||
FQ212702.1 | - | 2 - 3 | 190 | 112 | ||
FQ215129.1 | - | 2 - 3 | 200 | 112 | ||
FQ215603.1 | - | 2 - 3 | 185 | 112 | ||
FQ217383.1 | - | 2 - 3 | 190 | 112 | ||
FQ217819.1 | - | 190 | 112 | |||
FQ224891.1 | - | 2 - 3 | 188 | 112 | ||
FQ233582.1 | - | 2 - 3 | 190 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
