Assay Details
Gene Symbol: Mdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033235.1 | NP_150238.1 | 5 - 6 | 536 | 110 | |
GenBank mRNA | AF093773.1 | - | 5 - 6 | 536 | 110 | |
BC059124.1 | - | 5 - 6 | 577 | 110 | ||
FQ212702.1 | - | 5 - 6 | 580 | 110 | ||
FQ215129.1 | - | 5 - 6 | 590 | 111 | ||
FQ215603.1 | - | 5 - 6 | 575 | 110 | ||
FQ217383.1 | - | 5 - 6 | 580 | 110 | ||
FQ217819.1 | - | 580 | 110 | |||
FQ233582.1 | - | 5 - 6 | 580 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
