Assay Details
Gene Symbol: Ckb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012529.3 | NP_036661.3 | 65 | 99 | ||
GenBank mRNA | BC087656.1 | - | 64 | 99 | ||
BC127477.1 | - | 55 | 99 | |||
FM060701.1 | - | 70 | 99 | |||
M14400.1 | - | 77 | 98 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC127477.1, M14400.1, NM_012529.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
