Assay Details
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137633.1 | NP_001131105.1 | 9 - 10 | 1813 | 62 | |
XM_006240577.2 | XP_006240639.1 | 8 - 9 | 1821 | 62 | ||
GenBank mRNA | FQ212109.1 | - | 8 - 9 | 1571 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P507566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31163 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P507566 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P507566 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P507566 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P673011 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10887318 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887314 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887307 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887312 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887311 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887319 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887308 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887315 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887313 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887310 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887316 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887317 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887309 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10887306 | - | - |
Rat Genome 230 2.0 Array | 1388773_at | - | - |
Rat Genome 230 2.0 Array | 1388773_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381376_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375806_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375505_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375806_at | - | - |
Rat Genome 230 2.0 Array | 1381376_at | - | - |
Rat Genome 230 2.0 Array | 1375505_at | - | - |
Affymetrix HT Rat Focus Array | 1381376_at | - | - |
Affymetrix HT Rat Focus Array | 1388773_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375806_at | - | - |
Affymetrix HT Rat Focus Array | 1375505_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375806_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388773_at | - | - |
Affymetrix HT Rat Focus Array | 1375505_at | - | - |
Affymetrix HT Rat Focus Array | 1381376_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600011131.rn.1 | TC0600001591.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600011128.rn.1 | TC0600001591.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600011130.rn.1 | TC0600001591.rn.1 | RTA TC |
Rat Expression Set 230 | 1375505_PM_at | - | - |
Rat Expression Set 230 | 1388773_at | - | - |
Rat Expression Set 230 | 1375806_at | - | - |
Rat Expression Set 230 | 1375806_PM_at | - | - |
Rat Expression Set 230 | 1388773_PM_at | - | - |
Rat Expression Set 230 | 1375505_at | - | - |
Rat Expression Set 230 | 1381376_at | - | - |
Rat Expression Set 230 | 1381376_PM_at | - | - |
Rat Genome U34 Set | rc_AA963643_s_at | - | - |
Rat Genome U34 Set | rc_AI178392_at | - | - |
Rat Genome U34 Set | rc_AI145374_at | - | - |
Rat Genome U34 Set | rc_AI102578_at | - | - |
Rat Genome U34 Set | rc_AI111858_at | - | - |
Rat Genome U34 Set | rc_AI230786_at | - | - |
Rat Genome U34 Set | rc_AI103284_at | - | - |
Rat Genome U34 Set | rc_AI102578_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21960924 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369294 | - | - |
