Assay Details
Gene Symbol: Exoc3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106759.2 | NP_001100229.1 | 6 - 7 | 1672 | 74 | |
XM_006240572.2 | XP_006240634.1 | 6 - 7 | 1672 | 74 | ||
XM_006240573.2 | XP_006240635.1 | 6 - 7 | 1630 | 74 | ||
XM_006240574.2 | XP_006240636.1 | 6 - 7 | 4050 | 74 | ||
XM_006240575.2 | XP_006240637.1 | 6 - 7 | 1776 | 74 | ||
XM_006240576.2 | XP_006240638.1 | 6 - 7 | 1439 | 74 | ||
GenBank mRNA | BC166426.1 | - | 6 - 7 | 1672 | 74 | |
FQ230530.1 | - | 6 - 7 | 1691 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
