Assay Details
Gene Symbol: Exoc3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106759.2 | NP_001100229.1 | 8 - 9 | 1875 | 80 | |
XM_006240572.2 | XP_006240634.1 | 8 - 9 | 1875 | 80 | ||
XM_006240573.2 | XP_006240635.1 | 8 - 9 | 1833 | 80 | ||
XM_006240574.2 | XP_006240636.1 | 8 - 9 | 4253 | 80 | ||
XM_006240575.2 | XP_006240637.1 | 8 - 9 | 1979 | 80 | ||
XM_006240576.2 | XP_006240638.1 | 8 - 9 | 1642 | 80 | ||
GenBank mRNA | BC166426.1 | - | 8 - 9 | 1875 | 80 | |
FQ230530.1 | - | 8 - 9 | 1894 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
