Assay Details
Gene Symbol: Exoc3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106759.2 | NP_001100229.1 | 10 - 11 | 2148 | 93 | |
XM_006240572.2 | XP_006240634.1 | 10 - 11 | 2148 | 93 | ||
XM_006240573.2 | XP_006240635.1 | 10 - 11 | 2106 | 93 | ||
XM_006240574.2 | XP_006240636.1 | 10 - 11 | 4526 | 93 | ||
XM_006240575.2 | XP_006240637.1 | 10 - 11 | 2252 | 93 | ||
XM_006240576.2 | XP_006240638.1 | 10 - 11 | 1915 | 93 | ||
GenBank mRNA | BC166426.1 | - | 10 - 11 | 2148 | 93 | |
FQ230530.1 | - | 10 - 11 | 2167 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
