Assay Details
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013170.2 | NP_001013188.2 | 5 - 6 | 675 | 73 | |
XM_006240542.2 | XP_006240604.1 | 5 - 6 | 712 | 73 | ||
XM_008764811.1 | XP_008763033.1 | 5 - 6 | 866 | 73 | ||
XM_008764812.1 | XP_008763034.1 | 5 - 6 | 1463 | 73 | ||
GenBank mRNA | AY724504.1 | - | 5 - 6 | 675 | 73 | |
BC061752.1 | - | 6 - 7 | 738 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
