Assay Details
Gene Symbol: Begain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111115.1 | NP_001104585.1 | 5 - 6 | 441 | 71 | |
NM_024163.2 | NP_077077.1 | 5 - 6 | 461 | 71 | ||
XM_008764858.1 | XP_008763080.1 | 6 - 7 | 859 | 71 | ||
XM_008764859.1 | XP_008763081.1 | 5 - 6 | 769 | 71 | ||
XM_008764860.1 | XP_008763082.1 | 5 - 6 | 971 | 71 | ||
GenBank mRNA | AF064868.1 | - | 5 - 6 | 496 | 71 | |
AF064869.1 | - | 5 - 6 | 454 | 71 | ||
CB801805.1 | - | 1 - 2 | 180 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
