Assay Details
Gene Symbol: Mtrf1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025723.1 | NP_001020894.1 | 5 - 6 | 853 | 66 | |
GenBank mRNA | BC097970.1 | - | 5 - 6 | 853 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
