Assay Details
Gene Symbol: Nox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004216.1 | NP_001004216.1 | 5 - 6 | 489 | 68 | |
GenBank mRNA | AY573239.1 | - | 5 - 6 | 489 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
