Assay Details
Gene Symbol: Araf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033663.2 | NP_001028835.1 | 4 - 5 | 416 | 94 | |
NM_022532.2 | NP_071977.1 | 4 - 5 | 416 | 94 | ||
XM_008773036.1 | XP_008771258.1 | 4 - 5 | 1158 | 94 | ||
GenBank mRNA | AB158253.1 | - | 3 - 4 | 312 | 94 | |
BC060568.1 | - | 456 | 94 | |||
CO572305.1 | - | 4 - 5 | 416 | 94 | ||
X06942.1 | - | 3 - 4 | 383 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
