Assay Details
Gene Symbol: Srpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022524.1 | NP_071969.1 | 4 - 5 | 589 | 140 | |
XM_006256683.2 | XP_006256745.1 | 3 - 4 | 617 | 140 | ||
XM_008773077.1 | XP_008771299.1 | 3 - 4 | 1743 | 140 | ||
GenBank mRNA | BC087639.1 | - | 3 - 4 | 588 | 140 | |
D78359.1 | - | 4 - 5 | 589 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
