Assay Details
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106191.1 | NP_001099661.1 | 13 - 14 | 1720 | 87 | |
XM_006255751.2 | XP_006255813.1 | 13 - 14 | 1766 | 87 | ||
XM_006255752.2 | XP_006255814.1 | 13 - 14 | 1756 | 87 | ||
XM_006255753.2 | XP_006255815.1 | 12 - 13 | 1648 | 87 | ||
XM_006255754.2 | XP_006255816.1 | 12 - 13 | 1639 | 87 | ||
XM_006255755.2 | XP_006255817.1 | 12 - 13 | 1639 | 87 | ||
XM_006255756.2 | XP_006255818.1 | 12 - 13 | 1630 | 87 | ||
XM_006255757.2 | XP_006255819.1 | 11 - 12 | 1512 | 87 | ||
XM_008772601.1 | XP_008770823.1 | 13 - 14 | 1987 | 87 | ||
GenBank mRNA | BC160844.1 | - | 13 - 14 | 1747 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P082964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78860 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P082964 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P082964 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P082964 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P19045 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42627 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91094 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10808473 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808468 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808463 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808474 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808465 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808470 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808472 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808477 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808462 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808464 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808476 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808475 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808467 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808471 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808466 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808469 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10808461 | - | - |
Rat Genome 230 2.0 Array | 1383083_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383083_at | - | - |
Affymetrix HT Rat Focus Array | 1383083_at | - | - |
Affymetrix HT Rat Focus Array | 1383083_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900006223.rn.1 | TC1900000764.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900006222.rn.1 | TC1900000764.rn.1 | RTA TC |
Rat Expression Set 230 | 1383083_PM_at | - | - |
Rat Expression Set 230 | 1383083_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808461 | - | - |
Rat Genome U34 Set | rc_AI073109_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20991415 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352893 | - | - |
