Assay Details
Gene Symbol: Dpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053591.2 | NP_446043.1 | 7 - 8 | 918 | 114 | |
XM_006255783.2 | XP_006255845.1 | 7 - 8 | 913 | 114 | ||
GenBank mRNA | BC072476.1 | - | 7 - 8 | 918 | 114 | |
L07315.1 | - | 7 - 8 | 896 | 114 | ||
L07316.1 | - | 7 - 8 | 896 | 114 | ||
M94056.1 | - | 7 - 8 | 896 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
