Assay Details
Gene Symbol: Cdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031334.1 | NP_112624.1 | 14 - 15 | 2432 | 116 | |
GenBank mRNA | AB017696.1 | - | 14 - 15 | 2432 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
