Assay Details
Gene Symbol: Gab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108444.1 | NP_001101914.1 | 9 - 10 | 2206 | 126 | |
GenBank mRNA | GQ140625.1 | - | 10 - 11 | 2006 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
response to oxidative stress
signal transduction
epidermal growth factor receptor signaling pathway
heart development
epidermis development
regulation of cell migration
positive regulation of MAPK cascade
platelet-derived growth factor receptor signaling pathway
labyrinthine layer development
interleukin-6-mediated signaling pathway
response to oxidative stress
signal transduction
epidermal growth factor receptor signaling pathway
heart development
epidermis development
regulation of cell migration
positive regulation of MAPK cascade
platelet-derived growth factor receptor signaling pathway
labyrinthine layer development
interleukin-6-mediated signaling pathway
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P494608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49874 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P494608 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P494608 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P494608 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P494608 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90120 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53380 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52593 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10806933 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806935 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806936 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806930 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806932 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806928 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806931 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806934 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806929 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10806927 | - | - |
Rat Genome 230 2.0 Array | 1399167_a_at | - | - |
Rat Genome 230 2.0 Array | 1399167_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1399167_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1399167_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900003239.rn.1 | TC1900000408.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10806927 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20889210 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349894 | - | - |
