Assay Details
Gene Symbol: Pkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017175.2 | NP_058871.2 | 19 - 20 | 2624 | 123 | |
XM_006255245.2 | XP_006255307.1 | 19 - 20 | 3664 | 123 | ||
GenBank mRNA | BC061836.1 | - | 19 - 20 | 2525 | 123 | |
EV774518.1 | - | 2 - 3 | 244 | 123 | ||
FQ222217.1 | - | 19 - 20 | 2624 | 123 | ||
L35634.1 | - | 19 - 20 | 2467 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
