Assay Details
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011928.1 | NP_001011928.1 | 2 - 3 | 363 | 119 | |
XM_008772487.1 | XP_008770709.1 | 2 - 3 | 660 | 119 | ||
GenBank mRNA | BC088195.1 | - | 2 - 3 | 363 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
