Assay Details
Gene Symbol: Pdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107430.1 | NP_001100900.1 | 11 - 12 | 1653 | 124 | |
XM_006255596.2 | XP_006255658.2 | 13 - 14 | 1908 | 124 | ||
XM_008772514.1 | XP_008770736.1 | 12 - 13 | 1824 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
