Assay Details
Gene Symbol: Pdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107430.1 | NP_001100900.1 | 5 - 6 | 774 | 74 | |
XM_006255596.2 | XP_006255658.2 | 7 - 8 | 1029 | 74 | ||
XM_008772514.1 | XP_008770736.1 | 6 - 7 | 945 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
